Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900250

Sigma-Aldrich

2,4,6-Trimethylpyridine

Vetec, reagent grade, 98%

Synonym(s):

2,4,6-Collidine, sym-Collidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Beilstein/REAXYS Number:
107283
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.498 (lit.)

bp

171-172 °C (lit.)

mp

−43 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)nc(C)c1

InChI

1S/C8H11N/c1-6-4-7(2)9-8(3)5-6/h4-5H,1-3H3

InChI key

BWZVCCNYKMEVEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tissue fixative for electron microscopy.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Tomsig et al.
Histochemistry, 87(1), 21-25 (1987-01-01)
Rat pineal nerve endings contain a population of small and of large synaptic vesicles that are either electron lucent or have electron-dense cores. It has been reported that their osmiophilia is eliminated when collidine buffer is used in the fixation
Christa A Currie et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 824(1-2), 201-205 (2005-08-09)
Microchip capillary electrophoresis (CE), coupled with indirect fluorescence detection was investigated for estimating the pK(a) values of non-fluorescent compounds. The CE method is based on the differences in electrophoretic mobility of the analyte as a function of the pH of
G Rodríguez de Lores Arnaiz et al.
Neuroscience letters, 108(3), 309-313 (1990-01-22)
Previous histochemical studies have demonstrated that collidine abolished the osmiophilia and the chromaffin reaction of the synaptic vesicles, an effect which was ascribed to the release of stored neurotransmitters. Other studies indicated a relationship between the inhibition of Na+,K(+)-ATPase and
Jeffrey R Kanofsky et al.
Journal of photochemistry and photobiology. B, Biology, 87(2), 124-129 (2007-04-25)
Prior work using cell culture has shown that certain synthetic carotenoid derivatives are more effective than natural carotenoids in preventing photosensitized cell killing. These carotenoid derivatives are either cationic or are weak bases. We have now found that of one
Krisztina Sebok-Nagy et al.
Photochemistry and photobiology, 81(5), 1212-1218 (2005-05-20)
The fluorescent properties of 2-hydroxy Nile red dye (HONR) proved to be highly sensitive to the basicity of hydrogen bond acceptors. Fluorescence quantum yields and fluorescence decay profiles were measured as the function of the concentration of organic nitrogen compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service