Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900249

Sigma-Aldrich

Cyclohexanol

Vetec, reagent grade, 98%

Synonym(s):

1-Cyclohexanol, Cyclohexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein/REAXYS Number:
906744
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

product line

Vetec

assay

98%

form

solid or liquid

autoignition temp.

572 °F

expl. lim.

12.25 %

refractive index

n20/D 1.465 (lit.)

pH

6.5 (20 °C, 40 g/L)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohan Karande et al.
Biotechnology and bioengineering, 113(1), 52-61 (2015-07-15)
The applications of biocatalysts in chemical industries are characterized by activity, selectivity, and stability. One key strategy to achieve high biocatalytic activity is the identification of novel enzymes with kinetics optimized for organic synthesis by Nature. The isolation of novel
Matthew R Carstens et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), 8732-8737 (2015-07-01)
A new paradigm in oncology establishes a spectrum of tumorigenic potential across the heterogeneous phenotypes within a tumor. The cancer stem cell hypothesis postulates that a minute fraction of cells within a tumor, termed cancer stem cells (CSCs), have a
Xiao-Kun She et al.
Journal of chromatography. A, 1423, 31-38 (2015-11-13)
Porous lead(II)-based metal-organic nanotubes (Pb-MONTs) were, for the first time, used as an adsorbent for dispersive solid-phase extraction (d-SPE) of polybrominated diphenyl ethers (PBDEs) at trace levels from environmental water samples. Gas chromatography-negative chemical ionization-mass spectrometry (GC-NCI-MS) was applied for
Aamer Saeed et al.
Journal of fluorescence, 25(4), 1045-1053 (2015-06-19)
A series of dyes using tetrachloroperylene dianhydride as fluorescent chromophore was synthesized by the substitution of suitable aliphatic and alicyclic alcohols in alkaline medium, and evaluation of dyes was done for their optical, electrochemical and thermal properties. These dyes exhibited
Verena Resch et al.
AMB Express, 4, 30-30 (2014-06-21)
The Michael hydratase - alcohol dehydrogenase (MhyADH) from Alicycliphilus denitrificans was previously identified as a bi-functional enzyme performing a hydration of α,β-unsaturated ketones and subsequent oxidation of the formed alcohols. The investigations of the bi-functionality were based on a spectrophotometric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service