Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900248

Sigma-Aldrich

1,3-Dibromopropane

Vetec, reagent grade, 97.5%

Synonym(s):

Trimethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3Br
CAS Number:
Molecular Weight:
201.89
Beilstein/REAXYS Number:
635662
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

7 (vs air)

product line

Vetec

assay

97.5%

refractive index

n20/D 1.524 (lit.)

bp

167 °C (lit.)

mp

−34 °C (lit.)

density

1.989 g/mL at 25 °C (lit.)

SMILES string

BrCCCBr

InChI

1S/C3H6Br2/c4-2-1-3-5/h1-3H2

InChI key

VEFLKXRACNJHOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E E Badr et al.
Journal of oleo science, 59(12), 647-652 (2010-11-26)
A fatty hydrazide based cationic gemini surfactants, 1,3-bis (N'-acyl-N,N-dimethylhydrazinium) propane dibromide which possess hydrolyzable amido moieties in the lipophilic portions, were prepared by reacting 1,3-bromopropane with N,N-dimethyl fatty hydrazide. The surface properties were explained and discussed based on the effect
S P James et al.
Toxicology letters, 8(1-2), 7-15 (1981-04-01)
Oral administration of 1,3-dibromopropane (2 mmol/kg) to rats resulted in a marked decrease in the level of hepatic glutathione (GSH). Sulphur-containing [14C]-metabolites were excreted in the bile of rats dose with 1,3-bromo[14C]propane and were subjected to enterohepatic cycling. After an
A Witkowski et al.
The Journal of biological chemistry, 267(26), 18488-18492 (1992-09-15)
Thioesterase II is a 29-kDa monomer which, in certain specialized tissues, acts as a chain terminator in fatty acid synthesis by hydrolyzing medium-chain fatty acids from the fatty acid synthase. As with serine proteases, hydrolysis appears to involve acylation of
Sang Kyu Lee et al.
Journal of toxicology and environmental health. Part A, 70(15-16), 1381-1390 (2007-07-27)
To determine a possible role of glutathione (GSH) conjugation in 1,3-dibromopropane (1,3-DBP)-induced hepatotoxicity and immunotoxicity, female BALB/c mice were treated orally with 1,3-DBP. Based on the liquid chromatography/electrospray ionization-tandem mass spectrometry (LC/ESI-MS) analyses, two forms of S-bromopropyl GSH were observed
A R Jones et al.
Xenobiotica; the fate of foreign compounds in biological systems, 11(8), 541-546 (1981-08-01)
1. The metabolism of 1,3-dibromopropane had been investigated in the rat. Two conjugated metabolites have been isolated from the urine and identified as S-(3-hydroxypropyl)cysteine and N-acetyl-S-(3-hydroxypropyl)cysteine. 2. An oxidation product, identified as beta-bromolactic acid, has been isolated as a urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service