Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900243

Sigma-Aldrich

1,4-Butanediol

Vetec, reagent grade, 98%

Synonym(s):

1,4-Butylene glycol, Tetramethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1633445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
assay:
98%
bp:
230 °C (lit.)
Pricing and availability is not currently available.

grade

reagent grade

vapor density

3.1 (vs air)

product line

Vetec

assay

98%

autoignition temp.

698 °F

dilution

(for analytical testing)

refractive index

n20/D 1.445 (lit.)

bp

230 °C (lit.)

mp

16 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Rinne Dahl et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 885-886, 37-42 (2012-01-10)
The demand of high throughput methods for the determination of gamma-hydroxybutyrate (GHB) and its precursors gamma-butyrolactone (GBL) and 1,4-butane-diol (1,4BD) as well as for pregabalin is increasing. Here we present two analytical methods using ultra-high pressure liquid chromatography (UPLC) and
Anders Hamberg et al.
Chemical communications (Cambridge, England), 48(80), 10013-10015 (2012-09-05)
The enzyme Candida antarctica lipase B was subjected to site directed mutagenesis suggested by molecular modelling. The selectivity for the enzyme increased towards a range of diols over their corresponding monoesters as an effect of the mutations.
Ari Karchin et al.
Acta biomaterialia, 7(9), 3277-3284 (2011-06-07)
Electrospinning from a melt, in contrast to from a solution, is an attractive tissue engineering scaffold manufacturing process as it allows for the formation of small diameter fibers while eliminating potentially cytotoxic solvents. Despite this, there is a dearth of
Qin Zhou et al.
Metabolic engineering, 13(6), 777-785 (2011-10-26)
Conversion of 3-hydroxypropionate (3HP) from 1,3-propanediol (PDO) was improved by expressing dehydratase gene (dhaT) and aldehyde dehydrogenase gene (aldD) of Pseudomonas putida KT2442 under the promoter of phaCAB operon from Ralstonia eutropha H16. Expression of these genes in Aeromonas hydrophila
Linbo Wu et al.
Biomacromolecules, 13(9), 2973-2981 (2012-07-27)
Novel potentially biobased aliphatic-aromatic copolyesters poly(butylene succinate-co-butylene furandicarboxylate) (PBSFs) in full composition range were successfully synthesized from 2,5-furandicarboxylic acid (FA), succinic acid (SA), and 1,4-butanediol (BDO) via an esterification and polycondensation process using tetrabutyl titanate (TBT) or TBT/La(acac)(3) as catalyst.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service