Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900241

Sigma-Aldrich

Diethylene glycol

Vetec, reagent grade, 98%

Synonym(s):

2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein/REAXYS Number:
969209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
assay:
98%
bp:
245 °C (lit.)
vapor pressure:
0.01 mmHg ( 20 °C)

grade

reagent grade

vapor density

2.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

product line

Vetec

assay

98%

form

viscous liquid

autoignition temp.

442 °F

expl. lim.

2-12.3 %

dilution

(for analytical testing)

refractive index

n20/D 1.447 (lit.)

pH

5.0-8.0 (20 °C, 500 g/L)

bp

245 °C (lit.)

mp

−10 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

253.4 °F - closed cup

flash_point_c

123 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Handa et al.
Plant & cell physiology, 56(8), 1490-1511 (2015-05-27)
Gene expression during arbuscular mycorrhizal development is highly orchestrated in both plants and arbuscular mycorrhizal fungi. To elucidate the gene expression profiles of the symbiotic association, we performed a digital gene expression analysis of Lotus japonicus and Rhizophagus irregularis using
Pavel Kucheryavy et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 710-716 (2012-12-20)
Magnetite nanoparticles in the size range of 3.2-7.5 nm were synthesized in high yields under variable reaction conditions using high-temperature hydrolysis of the precursor iron(II) and iron(III) alkoxides in diethylene glycol solution. The average sizes of the particles were adjusted
Bichismita Sahu et al.
The Journal of organic chemistry, 76(14), 5614-5627 (2011-05-31)
Developed in the early 1990s, peptide nucleic acid (PNA) has emerged as a promising class of nucleic acid mimic because of its strong binding affinity and sequence selectivity toward DNA and RNA and resistance to enzymatic degradation by proteases and
Parisa Sadatmousavi et al.
Acta biomaterialia, 8(9), 3241-3250 (2012-05-30)
Self-assembling peptide nanofibers have emerged as important nanobiomaterials, with such applications as delivery of therapeutic agents and vaccines, nanofabrication and biomineralization, tissue engineering and regenerative medicine. Recently a new class of self-assembling peptides has been introduced, which takes into consideration
Asuman C Kucuk et al.
Langmuir : the ACS journal of surfaces and colloids, 27(10), 6381-6388 (2011-04-16)
"Core-corona" type amphiphiles, which comprise double-decker-shaped POSSs (DDSQs) as the core and two or four di(ethylene glycol) (DEG) units as the coronae, have recently been reported to form a stable monolayer at the air-water interface. In this paper, another core-corona

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service