Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900237

Sigma-Aldrich

Diethylene glycol dimethyl ether

Vetec, reagent grade, 98%

Synonym(s):

2-Methoxyethyl ether, Bis(2-methoxyethyl) ether, Dimethyldiglycol, ‘Diglyme’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2O
CAS Number:
Molecular Weight:
134.17
Beilstein/REAXYS Number:
1736101
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
grade:
reagent grade
assay:
98%
bp:
162 °C (lit.)
vapor pressure:
3 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.6 (vs air)

vapor pressure

3 mmHg ( 20 °C)

product line

Vetec

assay

98%

form

liquid

autoignition temp.

370 °F

expl. lim.

17.4 %

dilution

(for analytical testing)

refractive index

n20/D 1.408 (lit.)

bp

162 °C (lit.)

mp

−64 °C (lit.)

density

0.944 g/mL at 20 °C (lit.)
0.939 g/mL at 25 °C (lit.)

SMILES string

COCCOCCOC

InChI

1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3

InChI key

SBZXBUIDTXKZTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Flam. Liq. 3 - Repr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

134.6 °F - closed cup

flash_point_c

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Catalano
Statistics in medicine, 16(8), 883-900 (1997-04-30)
Simultaneous observation of continuous and ordered categorical outcomes for each subject is common in biomedical research but multivariate analysis of the data is complicated by the multiple data types. Here we construct a model for the joint distribution of bivariate
Martin Schulz-Dobrick et al.
Journal of the American Chemical Society, 127(37), 12816-12817 (2005-09-15)
A new, facile and generally applicable synthesis of functionalized gold nanoparticles is presented. It is based on the surfactant-free generation of weakly stabilized nanoparticles by the reduction of HAuCl4 with sodium naphthalenide in diglyme. These nanoparticles were found to lack
Adriana Valore et al.
Journal of the American Chemical Society, 132(13), 4966-4970 (2010-03-17)
The second-order nonlinear optical (NLO) properties of [Ln(hfac)(3)(diglyme)] (hfac = hexafluoroacetylacetonate; diglyme = bis(2-methoxyethyl)ether; Ln = La, Ce, Pr, Sm, Eu, Gd, Er, Lu) complexes have been investigated by a combination of electric-field second harmonic generation (EFISH) and harmonic light
Patrik Johansson et al.
The journal of physical chemistry. A, 114(39), 10700-10705 (2010-09-11)
The conformational states of diglyme in both the solid and liquid states have been determined by an integrated use of both IR and Raman spectroscopy together with ab initio/density functional theory (DFT) calculations. Using knowledge from diglyme also made possible
P J Catalano et al.
Teratology, 47(4), 281-290 (1993-04-01)
The purpose of this paper is to present a statistical model for analyzing the joint effects of exposure on fetal death, fetal weight, and malformation in a developmental toxicity study. In addition to allowing for the usual litter effect, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service