Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900235

Sigma-Aldrich

Diethylene glycol diethyl ether

Vetec, reagent grade, 98+ %

Synonym(s):

2-Ethoxyethyl ether, Bis(2-ethoxyethyl) ether, Diethyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5OCH2CH2)2O
CAS Number:
Molecular Weight:
162.23
Beilstein/REAXYS Number:
1699259
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.6 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)

product line

Vetec

assay

≥98%

form

liquid

expl. lim.

6.2 % (lit.)

refractive index

n20/D 1.412 (lit.)

bp

180-190 °C (lit.)

mp

-68 °C (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCOCC

InChI

1S/C8H18O3/c1-3-9-5-7-11-8-6-10-4-2/h3-8H2,1-2H3

InChI key

RRQYJINTUHWNHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lianfang Chen et al.
Journal of chromatography. A, 1394, 103-110 (2015-04-09)
A novel organic monolith was firstly prepared in a UV-transparent fused-silica capillary by a single-step approach via photo-initiated thiol-ene click polymerization reaction of 1,2,4-trivinylcyclohexane (TVCH) and pentaerythriol tetra(3-mercaptopropionate) (4SH) within 10min. The effects of both composition of prepolymerization solution and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service