Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900225

Sigma-Aldrich

Benzyl benzoate

Vetec, reagent grade, 98%

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein/REAXYS Number:
2049280
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

1 mmHg ( 125 °C)

product line

Vetec

assay

98%

autoignition temp.

896 °F

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

298.4 °F - closed cup

flash_point_c

148 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine W McCollum et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 152-163 (2014-04-29)
Exposure to arsenic in its inorganic form, arsenite, causes adverse effects to many different organs and tissues. Here, we have investigated arsenite-induced adverse effects on vascular tissues in the model organism zebrafish, Danio rerio. Zebrafish embryos were exposed to arsenite
Karina Caballero-Gallardo et al.
Journal of agricultural and food chemistry, 59(5), 1690-1696 (2011-02-05)
A tool for integrated pest management is the use of essential oils (EOs) and plant extracts. In this study, EOs from Tagetes lucida , Lepechinia betonicifolia , Lippia alba , Cananga odorata , and Rosmarinus officinalis , species grown in
Fatimata Ly et al.
Bulletin of the World Health Organization, 87(6), 424-430 (2009-07-01)
To compare the effectiveness of oral ivermectin (IV) and two different modalities of topical benzyl benzoate (BB) for treating scabies in a community setting. The trial included patients aged 5-65 years with scabies who attended the dermatology department at the
Birke Walter et al.
Archives of dermatology, 147(4), 468-473 (2011-04-13)
Scabies is a parasitic skin disease endemic in resource-poor communities in low-income countries. The best ways to diagnose scabies in this setting have not been investigated. To compare the diagnostic properties of dermoscopy, the microscopic examination of a skin scraping
Osamu Ohno et al.
Bioorganic & medicinal chemistry, 16(16), 7843-7852 (2008-08-02)
Hypertension is a lifestyle-related disease which often leads to serious conditions such as heart disease and cerebral hemorrhage. Angiotensin II (Ang II) plays an important role in regulating cardiovascular homeostasis. Consequently, antagonists that block the interaction of Ang II with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service