Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900223

Sigma-Aldrich

Ammonium bromide

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Br
CAS Number:
Molecular Weight:
97.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

vapor pressure

1 mmHg ( 198.3 °C)

product line

Vetec

assay

98%

form

powder or crystals

pH

5-6

mp

452 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

SMILES string

N.Br[H]

InChI

1S/BrH.H3N/h1H;1H3

InChI key

SWLVFNYSXGMGBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Repr. 1B - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirk J Ziegler et al.
Nano letters, 5(12), 2355-2359 (2005-12-15)
A two-phase liquid-liquid extraction process is presented which is capable of extracting water-soluble single-walled carbon nanotubes into an organic phase. The extraction utilizes electrostatic interactions between a common phase transfer agent and the sidewall functional groups on the nanotubes. Large
Lei Cheng et al.
Dental materials : official publication of the Academy of Dental Materials, 28(8), 853-862 (2012-05-15)
Half of dental restorations fail in 10 years, with secondary caries as the main reason. Calcium phosphate composites could remineralize tooth lesions. The objectives of this study were to: (1) impart antibacterial activity to a composite with nanoparticles of amorphous
Catalytic synthesis of nonracemic azaproline derivatives by cyclization of β-alkynyl hydrazines under kinetic resolution conditions.
Pradip Maity et al.
Angewandte Chemie (International ed. in English), 50(36), 8338-8341 (2011-07-19)
Fredrik Hallberg et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 192(1), 69-77 (2008-02-22)
Although simple as a concept, electrophoretic NMR (eNMR) has so far failed to find wider application. Problems encountered are mainly due to disturbing and partly irreproducible convection-like bulk flow effects from both electro-osmosis and thermal convection. Additionally, bubble formation at
Erhong Duan et al.
Journal of hazardous materials, 194, 48-52 (2011-08-27)
The solubility of SO(2) in a binary mixture of water and caprolactam tetrabutyl ammonium bromide ionic liquid (CPL-TBAB IL) was investigated. Though the ionic liquid and water were fully miscible, a phase separation occurred when SO(2) was introduced into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service