Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900220

Sigma-Aldrich

Sulfanilic acid

Vetec, reagent grade, 99%

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein/REAXYS Number:
908765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

powder or crystals

mp

>300 °C (lit.)

solubility

5% Na2CO3 solution: 0.1 mg/mL, clear, colorless to almost colorless

density

1.4862 g/cm3 at 20 °C (OECD Test Guideline 109)

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam Hoon Kim et al.
Nano letters, 10(10), 4181-4185 (2010-09-25)
A bifunctional adenosine-sensitive double-stranded DNA aptamer was used to create and control a surface-enhanced Raman spectroscopy (SERS) hot spot between a bulk Au surface and a gold nanoparticle (Au NP) attached to the aptamer via a biotin-avidin linkage. The Au
Reversible tuning of SERS hot spots with aptamers.
Nam Hoon Kim et al.
Advanced materials (Deerfield Beach, Fla.), 23(36), 4152-4156 (2011-08-09)
Nam-Jung Kim et al.
Chemical communications (Cambridge, England), (41)(41), 6246-6248 (2009-10-15)
An elusive chemical contribution to SERS is investigated by estimating the chemical coupling strengths through a dynamic relative peak analysis during solvent evaporation from a diluted chemical sample that is drop-deposited on a soft SERS substrate.
Prashant Barsing et al.
Bioresource technology, 102(2), 765-771 (2010-09-25)
A novel bacterial consortium (TJ-2) for mineralization of aromatic amines resulting from decolorization of azo dyes was developed. Three bacterial strains were identified as Pseudomonas pseudoalcaligenes (TJ-21,EU072476), Pseudomonas citronellolis (TJ-22,EU072477) and Pseudomonas testosterone (TJ-23,EU072477) by 16S rRNA gene sequence analysis.
Jinghua Chen et al.
Analytical chemistry, 80(21), 8028-8034 (2008-09-26)
In this study, an electrochemical DNA biosensor was developed for detection of the breakpoint cluster region gene and the cellular abl (BCR/ABL) fusion gene in chronic myelogenous leukemia by using 18-mer locked, nucleic acid-modified, single-stranded DNA as the capture probe.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service