Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900219

Sigma-Aldrich

Barium hydroxide octahydrate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · 8H2O
CAS Number:
Molecular Weight:
315.46
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

pH

12.5 (20 °C, 50 g/L)

mp

78 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O[Ba]O

InChI

1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2

InChI key

ZUDYPQRUOYEARG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyzes the β-elimination of phosphoserine residues.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Verhaeghe et al.
Clinical chemistry, 34(6), 1077-1083 (1988-06-01)
This is a new method for isolating and concentrating urinary organic acids before gas chromatography--mass spectrometry. Sulfate and phosphate anions are removed by precipitation with Ba(OH)2, and the urine pH is adjusted to 8-8.5. The sample is then applied onto
L Toniolo et al.
Annali di chimica, 91(11-12), 813-821 (2002-02-12)
The Arch of Peace, by Luigi Cagnola, is one of the most famous neoclassical monuments in Milan. It has been subjected to conservative intervention in 1998. In the present paper the efficacy of the consolidation by means of barium hydroxide
K Freeman et al.
Journal of endodontics, 20(6), 272-275 (1994-06-01)
To study the ability of calcium hydroxide to promote hard tissue repair, Alza Alzet Osmotic Pumps, implanted in Sprague-Dawley rats, were used to deliver either calcium hydroxide and glycerol, barium hydroxide and glycerol, tetracycline and glycerol, or glycerol only to
A Mchaourab et al.
Anesthesiology, 94(6), 1007-1009 (2001-07-24)
Potent inhaled anesthetics degrade in the presence of the strong bases (sodium hydroxide or potassium hydroxide) in carbon dioxide (CO2) absorbents. A new absorbent, Amsorb (Armstrong Medical Ltd., Coleraine, Northern Ireland), does not employ these strong bases. This study compared
V V Onoprienko et al.
Bioorganicheskaia khimiia, 26(6), 403-410 (2000-08-03)
Peptides Boc-Ala-Asn/Gln-OH and Boc-Asn/Gln-Ala-OH were saponified with barium hydroxide to corresponding Asp/Glu-containing peptides. Under the conditions of saponification, Boc-Asn-Ala-OH additionally afforded Boc-Asp-OH, isopeptide Boc-Asp(Ala)-OH, and Boc-NHSuc > Ala-OH, with the third being the key intermediate in these transformations. Boc-Asp(OMe)-Ala-OMe underwent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service