Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900215

Sigma-Aldrich

2-Octanol

Vetec, reagent grade, 97%

Synonym(s):

sec-Caprylic alcohol, Capryl alcohol, Ethyl-N-amylcarbinol, Ethylpentylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
reagent grade
assay:
97%
bp:
174-181 °C (lit.)
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

dilution

(for analytical testing)

refractive index

n20/D 1.426 (lit.)

bp

174-181 °C (lit.)

mp

-38 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Rees et al.
Biochimica et biophysica acta, 1073(3), 493-501 (1991-04-09)
Lipase from three different sources has been immobilised in microemulsion-based gels (MBGs) with retention of catalytic activity. Such lipase-containing MBGs prove to be novel solid-phase catalysts for use in apolar organic solvents such as n-heptane. Using these systems, preparative-scale synthesis
Chi-Yu Huang et al.
International journal of pharmaceutics, 325(1-2), 132-139 (2006-07-22)
Core-shell type of nanoparticles (NPs) with manipulated degradation rate and balanced hydrophilic/hydrophobic properties were designed and characterized. The NPs based on the copolymers of n-butyl cyanoacrylate (BCA) and 2-octyl cyanoacrylate (OCA) were prepared by anion emulsion polymerization in 0.01N HCl
J S Schweitzer et al.
Journal of neurophysiology, 84(2), 927-933 (2000-08-12)
Under conditions of low [Ca(2+)](o) and high [K(+)](o), the rat dentate granule cell layer in vitro develops recurrent spontaneous prolonged field bursts that resemble an in vivo phenomenon called maximal dentate activation. To understand how pH changes in vivo might
Deracemization of secondary alcohols through a concurrent tandem biocatalytic oxidation and reduction.
Constance V Voss et al.
Angewandte Chemie (International ed. in English), 47(4), 741-745 (2007-12-12)
Er-na Xun et al.
Applied biochemistry and biotechnology, 168(3), 697-707 (2012-08-01)
The lipase from Pseudomonas fluorescens (Lipase AK, AKL) was immobilized onto the magnetic Fe(3)O(4) nanoparticles via hydrophobic interaction. Enzyme loading and immobilization yield were determined as 21.4±0.5 mg/g and 49.2±1.8 %, respectively. The immobilized AKL was successfully used for resolution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service