Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900206

Sigma-Aldrich

Calcium oxide

Vetec, reagent grade

Synonym(s):

Lime, Quicklime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

≥68% Ca (EDTA titration)

pH

12.6 (20 °C)

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

SMILES string

O=[Ca]

InChI

1S/Ca.O

InChI key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Nazaret González-Alcaraz et al.
Journal of environmental management, 116, 72-80 (2013-01-08)
The aim of this study was to identify the effectiveness of liming in combination with vegetation for the recovery of slightly acidic, saline soils of eutrophic wetlands affected by mine wastes, under fluctuating flooding conditions. Simulated soil profiles were constructed
Sebastião Roberto Soares et al.
Waste management (New York, N.Y.), 33(1), 175-183 (2012-11-06)
The establishment of rules to manage Health Care Waste (HCW) is a challenge for the public sector. Regulatory agencies must ensure the safety of waste management alternatives for two very different profiles of generators: (1) hospitals, which concentrate the production
Vasilije Manovic et al.
International journal of environmental research and public health, 7(8), 3129-3140 (2010-10-16)
This paper presents a review of the research on CO(2) capture by lime-based looping cycles undertaken at CanmetENERGY's (Ottawa, Canada) research laboratories. This is a new and very promising technology that may help in mitigation of global warming and climate
Kunlun Zhang et al.
Journal of hazardous materials, 243, 278-285 (2012-11-20)
Tetrabromobisphenol A (TBBPA) is the most widely used brominated flame retardant (BFR), which has received more and more concerns due to its high lipophilicity, persistency and endocrine disrupting property in the environment. Considering the possible need for the safe disposal
Arup Roy et al.
Journal of biomedical nanotechnology, 9(9), 1570-1578 (2013-08-29)
The high degree of microbial diseases and their multidrug resistant properties make the researchers to develop new class of antimicrobial agents. A modern and innovative approach of drug development is the use of metallic nanoparticles as new formulations of antimicrobial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service