Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900204

Sigma-Aldrich

Magnesium hydroxide

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
39.4-44.0% Mg basis (EDTA titration)
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

39.4-44.0% Mg basis (EDTA titration)

form

powder

concentration

39.4-44.0% Mg (EDTA titration)

pH

9.5-10.5

mp

350 °C (lit.)

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francys K V Moreira et al.
Carbohydrate polymers, 92(2), 1743-1751 (2013-02-13)
In this paper the mechanical reinforcement of nano-sized brucite, Mg(OH)(2) in a series of bionanocomposite films based on starch was investigated. Brucite nanoplates with an aspect ratio of 9.25 were synthesized by wet precipitation and incorporated into starch matrices at
Xiaohong Pan et al.
ACS applied materials & interfaces, 5(3), 1137-1142 (2013-01-11)
Here we reported the antibacterial effect and related mechanism of three nano-Mg(OH)(2) slurries using Escherichia coli as model bacteria. X-ray diffraction (XRD), scanning electron microscopy (SEM) and laser particle size analysis revealed that the as-synthesized Mg(OH)(2)_(MgCl2), Mg(OH)(2)_(MgSO4) and Mg(OH)(2)_(MgO) are
Mayuko Y Kumasaka et al.
Archives of toxicology, 87(3), 439-447 (2012-10-27)
Various carcinomas including skin cancer are explosively increasing in arsenicosis patients who drink arsenic-polluted well water, especially in Bangladesh. Although well drinking water in the cancer-prone areas contains various elements, very little is known about the effects of elements except
Jia Zhang et al.
Journal of hazardous materials, 239-240, 128-134 (2012-09-19)
This paper describes the enhanced Cr(VI)-contaminated soil remediation via a combination of electrokinetics (EK) with a calcined-hydrotalcite-based permeable reactive barrier (PRB). First, this combination proved to be feasible, and remarkably facilitated Cr(VI) remediation in a column test. Then, lightly-to-severely (0.16-1.65
Suresh Kumar Kailasa et al.
The Analyst, 137(19), 4490-4496 (2012-08-11)
We report a dispersive liquid-liquid microextraction (LLME) method using surface modified Mg(OH)(2) nanoparticles (NPs) with oleic acid (OA) as hydrophobic affinity probes for the extraction and preconcentration of hydrophobic biomolecules prior to MALDI MS analysis. OA-capped Mg(OH)(2) NPs were characterized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service