Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900196

Sigma-Aldrich

Titanium(IV) oxide

Vetec, reagent grade, 99%

Synonym(s):

Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

nanoparticles

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

cation traces

As: ≤0.001%
Fe: ≤0.005%
Pb: ≤0.005%

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suxin Gui et al.
Journal of agricultural and food chemistry, 61(37), 8959-8968 (2013-08-24)
TiO₂ nanoparticles (NPs) are used in the food industry but have potential toxic effects in humans and animals. TiO₂ NPs impair renal function and cause oxidative stress and renal inflammation in mice, associated with inhibition of nuclear factor erythroid-2-related factor
D Minetto et al.
Environment international, 66, 18-27 (2014-02-11)
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:
Elisa Moschini et al.
Toxicology letters, 222(2), 102-116 (2013-08-03)
Metal oxide NPs are abundantly produced in nanotech industries and are emitted in several combustion processes, suggesting the need to characterize their toxic impact on the human respiratory system. The acute toxicity and the morphological changes induced by copper oxide
Susan C Tilton et al.
Nanotoxicology, 8(5), 533-548 (2013-05-11)
The growing use of engineered nanoparticles (NPs) in commercial and medical applications raises the urgent need for tools that can predict NP toxicity. Global transcriptome and proteome analyses were conducted on three human cell types, exposed to two high aspect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service