Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900195

Sigma-Aldrich

Sodium pyrophosphate decahydrate

Vetec, reagent grade, 99%

Synonym(s):

Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99%
grade:
reagent grade
form:
crystals or chunks
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

crystals or chunks

pH

9.5-10.5 (20 °C, 50 g/L)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric A Perras et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 211(2), 234-239 (2011-06-10)
Double-rotation (DOR) is the only technique generally capable of yielding high-resolution NMR spectra of half-integer quadrupolar nuclei in one dimension for solids without the need for sophisticated coherence pathway selection. Unfortunately, due to the low outer rotor spinning frequencies currently
Matthew Prideaux et al.
PloS one, 7(5), e36786-e36786 (2012-05-16)
Osteocytes are terminally differentiated osteoblasts which reside in a mineralized extracellular matrix (ECM). The factors that regulate this differentiation process are unknown. We have investigated whether ECM mineralization could promote osteocyte formation. To do this we have utilised MLO-A5 pre-osteocyte-like
Sanko Nguyen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 75-83 (2010-09-30)
The potential of liposomes as a drug delivery system for use in the oral cavity has been investigated. Specifically targeting for the teeth, the in vitro adsorption of charged liposomal formulations to hydroxyapatite (HA), a common model substance for the
Taís Scaramucci et al.
Journal of dentistry, 39(12), 841-848 (2011-09-29)
To evaluate the erosive potential of orange juice modified by food additives in enamel and dentine. Calcium lactate pentahydrate (CLP), xanthan gum (XG), sodium linear polyphosphate (LPP), sodium pyrophosphate tetrabasic (PP), sodium tripolyphosphate (STP) and some of their combinations were
Priyadip Das et al.
Chemical communications (Cambridge, England), 47(28), 8118-8120 (2011-06-21)
Newly synthesized Zn(II) and Cd(II)-based complexes show unique selectivity towards inorganic pyrophosphate (PPi) in 100% aqueous medium at pH = 7.4 and act as a "turn-on" and "turn-off" real-time assay, respectively, for the evaluation of the enzymatic activity of alkaline

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service