Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900187

Sigma-Aldrich

Malonic acid

Vetec, reagent grade, 98%

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein/REAXYS Number:
1751370
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

chunks
powder, crystals or granules

mp

132-135 °C (dec.) (lit.)

solubility

1 M NaOH: 100 mg/mL, clear to hazy, colorless to faintly yellow

density

1.619 g/cm3 at 25 °C

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Chand Ali et al.
Talanta, 203, 16-22 (2019-06-17)
The efficient extraction of natural products from biomass is deemed an essential area of interest using green technology in the pharmaceutical and biochemical industries. In recent years, deep eutectic solvents (DESs) have been paying broad interest as compatible, safe, and
E A Haan et al.
European journal of pediatrics, 144(6), 567-570 (1986-04-01)
A second child with a more severe deficiency of malonyl CoA decarboxylase is described. He is mildly mentally retarded and presented with vomiting, a seizure, hypoglycaemia and mild metabolic acidosis during a urinary tract infection. The urine contained increased amounts
Bogdan Belgorodsky et al.
Bioconjugate chemistry, 16(5), 1058-1062 (2005-09-22)
The preparation and characterization of the stable human serum albumin (HSA)-C3 isomer of tris-malonic acid [C60]fullerene complex is reported. Other than the anti-fullerene antibody, a stable protein-fullerene complex with a native protein has never been observed. This study may provide
Christian Bonhomme et al.
Journal of the American Chemical Society, 134(30), 12611-12628 (2012-06-29)
Strontium is an element of fundamental importance in biomedical science. Indeed, it has been demonstrated that Sr(2+) ions can promote bone growth and inhibit bone resorption. Thus, the oral administration of Sr-containing medications has been used clinically to prevent osteoporosis
Adriana Wilhelm et al.
Journal of medicinal chemistry, 55(22), 9817-9830 (2012-10-31)
The protein kinase C-related kinase 2 (PRK2)-interacting fragment (PIF) pocket of phosphoinositide-dependent kinase-1 (PDK1) was proposed as a novel target site for allosteric modulators. In the present work, we describe the design, synthesis, and structure-activity relationship of a series of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service