Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900184

Sigma-Aldrich

Sodium tetraphenylborate

Vetec, reagent grade, 98%

Synonym(s):

Tetraphenylboron sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4BNa
CAS Number:
Molecular Weight:
342.22
Beilstein/REAXYS Number:
3599783
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
98%
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

SMILES string

[Na+].c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.Na/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;/q-1;+1

InChI key

HFSRCEJMTLMDLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirela Samardžić et al.
Talanta, 83(3), 789-794 (2010-12-15)
A sensitive potentiometric surfactant sensor based on a highly lipophilic 1,3-didecyl-2-methyl-imidazolium cation and a tetraphenylborate (TPB) antagonist ion was used as the end-point detector in ion-pair potentiometric surfactant titrations using sodium TPB as a titrant. Several analytical and technical grade
Hiroyuki Hayashi et al.
The Journal of organic chemistry, 73(22), 8683-8693 (2008-10-16)
1-Ethyl-2-(1-oxy-3-oxo-4,4,5,5-tetramethylimidazolin-2-yl)-3-methylimidazolium bromide, [EMINN](+)[Br](-), carrying nitronylnitroxide (NN) in the cation unit, was prepared as a parent molecule and converted to seven salts, [EMINN](+)[X](-) (X = I, TFSI (bis(trifluoromethanesulfonyl)imide), BPh4 (tetraphenylborate), [EMINN](+)(1-3)[BA(1-3)]((1-3)-); BA1 (benzoic acid), BA2 (terephthalic acid), and BA3 (trimesic acid)
Beata Paczosa-Bator et al.
Talanta, 81(3), 1003-1009 (2010-03-20)
This paper focuses on the quantitative determination of the loss of the components from plastic membranes of ion-selective electrodes (ISEs) during contact with aqueous bathing solutions. The leaching processes, which affect the ISE responses, are rarely characterized by independent methods.
D C Kapsimali et al.
Talanta, 80(3), 1311-1317 (2009-12-17)
Two different derivatizing reagents were tested for the development of a fast and sensitive method for the determination of selenites (Se(IV)) in human urine. The reagents were sodium tetraethylborate (NaBEt(4)) and tetraphenylborate (NaBPh(4)), respectively, and the procedure is based on
M M Khater et al.
Bioelectrochemistry (Amsterdam, Netherlands), 77(1), 53-59 (2009-08-18)
In this study a PVC membrane electrode for determination of ketotifen fumarate is reported, where ketotifen tetraphenylborate (Keto-TPB) was used as ion exchanger. The electrode has linear range of 5.6x10(-6)-1.0x10(-2) and 1.0x10(-5)-1.0x10(-2) mol/L, with detection limits 2.37x10(-6)and 4.60x10(-6) mol/L in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service