V900181
Potassium hexacyanoferrate(II) trihydrate
Vetec™, reagent grade, 98%
Synonym(s):
Potassium ferrocyanide, Yellow prussiate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
K4Fe(CN)6 · 3H2O
CAS Number:
Molecular Weight:
422.39
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade
reagent grade
product line
Vetec™
assay
98%
form
powder
pH
8.0-10 (25 °C, 211 g/L)
mp
70 °C (lit.)
SMILES string
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N
InChI
1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;
InChI key
NRYOVBQTVIWDNU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
supp_hazards
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marta Mesías et al.
Food & function, 10(10), 6624-6632 (2019-09-26)
The biscuit industry has evolved in the last decades with the inclusion of alternative cereals, pseudo-cereals and other ingredients in traditional recipes, which allows current consumers' needs to be met. The dough composition, thermal treatment applied during baking, low moisture
Francisco J Morales et al.
Nutrients, 12(5) (2020-05-20)
Nutritional composition and neo-formed contaminant content in ultra-processed foods, amongst other factors, may contribute to increasing overall risk of non-communicable diseases and cancer. Commercial breakfast cereals (n = 53) were classified according to the NOVA approach as un-/minimally processed (NOVA-1
Marta Mesias et al.
Food & function, 11(2), 1489-1497 (2020-01-29)
Children are one of the most exposed groups to dietary acrylamide with 'potato fried products' that account for up to half the total exposure to this contaminant. Acrylamide was measured in French fries prepared in 31 primary school canteens randomly
C M Gabardo et al.
The Analyst, 140(16), 5781-5788 (2015-07-17)
Fully-integrated electro-fluidic systems with micro-/nano-scale features have a wide range of applications in lab-on-a-chip systems used for biosensing, biological sample processing, and environmental monitoring. Rapid prototyping of application-specific electro-fluidic systems is envisioned to facilitate the testing, validation, and market translation
Concha Bosch-Navarro et al.
Physical chemistry chemical physics : PCCP, 17(44), 29628-29636 (2015-10-20)
The electrochemical activity of graphene is of fundamental importance to applications from energy storage to sensing, but has proved difficult to unambiguously determine due to the challenges innate to fabricating well defined graphene electrodes free from contamination. Here, we report
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service