Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900177

Sigma-Aldrich

Aluminum fluoride trihydrate

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlF3·3H2O
CAS Number:
Molecular Weight:
138.02
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

≥18.9% (EDTA titration)

density

2.88 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].F[Al](F)F

InChI

1S/Al.3FH.3H2O/h;3*1H;3*1H2/q+3;;;;;;/p-3

InChI key

BKLZIAYVINRQEJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service