Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900174

Sigma-Aldrich

Calcium stearate

Vetec, reagent grade

Synonym(s):

Octadecanoic acid calcium salt, Stearic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COO]2Ca
CAS Number:
Molecular Weight:
607.02
Beilstein/REAXYS Number:
3919698
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Ca]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Ca/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

CJZGTCYPCWQAJB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service