Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900173

Sigma-Aldrich

Tetrabutylammonium bromide

Vetec, reagent grade, 99%

Synonym(s):

N,N,N-tributyl-1-butanaminium bromide, TBAB, TBABr, tetra-n-butylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(Br)
CAS Number:
Molecular Weight:
322.37
Beilstein/REAXYS Number:
3570983
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
99%
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

102-106 °C (lit.)

SMILES string

[Br-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

Inchi Key

JRMUNVKIHCOMHV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanumantha Rao Vutukuri et al.
Angewandte Chemie (International ed. in English), 53(50), 13830-13834 (2014-11-05)
Particle shape is a critical parameter that plays an important role in self-assembly, for example, in designing targeted complex structures with desired properties. Over the last decades, an unprecedented range of monodisperse nanoparticle systems with control over the shape of
Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind
Toshihiro Kimura et al.
Inorganic chemistry, 51(23), 13001-13008 (2012-11-15)
A simple monomeric tungstate, TBA(2)[WO(4)] (I, TBA = tetra-n-butylammonium), could act as an efficient homogeneous catalyst for chemical fixation of CO(2) with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Various kinds of structurally diverse 2-aminobenzonitriles could be converted into the corresponding quinazoline-2,4(1H,3H)-diones in high
R Bryan Sears et al.
Journal of inorganic biochemistry, 121, 77-87 (2013-01-29)
The complex cis-[Ru(phpy)(phen)(CH3CN)2](+) (phpy=2-phenylpyridine, phen=1,10-phenanthroline) was investigated as a potential photodynamic therapy (PDT) agent. This complex presents desirable photochemical characteristics including a low energy absorption tail extending into the PDT window (600-850nm) and photoinduced exchange of the CH3CN ligands, generating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service