Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900171

Sigma-Aldrich

Potassium disulfite

Vetec, reagent grade, 98%

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

supp_hazards

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Rojo-Bezares et al.
International journal of food microbiology, 116(1), 32-36 (2007-02-27)
Nisin is a bacteriocin used against food spoilage bacteria. Sulphur dioxide is a potent antioxidant as well as an antimicrobial agent widely used in the wine industry. In this study we describe the effect of these important antibacterial agents on
M Kaul et al.
Folia microbiologica, 41(2), 159-164 (1996-01-01)
The effectiveness of two commonly used preservatives, sodium benzoate and potassium disulfite, was evaluated in terms of their bactericidal activity and capacity to induce changes in the surface properties of Escherichia coli isolated from commercial food preserves. Preservative treatment over
M K Dutt
Microscopica acta, 84(3), 245-248 (1981-05-01)
This communication presents a method for the preparation of Schiff reagent in which N hydrochloric acid has been replaced by a low concentration of propionic acid. The result of using such a Schiff reagent indicates that the dye-reagent thus prepared
G Spano et al.
Current microbiology, 54(1), 9-13 (2006-12-13)
In order to monitor Lactobacillus plantarum and Oenococcus oeni in red wine produced with Italian grape (variety "Primitivo di Puglia"), a polymerase chain reaction- denaturing gradient gel electrophoresis (PCR-DGGE) approach using the rpoB as gene target was established. Wine was
D A Mathison et al.
Chest, 87(1 Suppl), 50S-54S (1985-01-01)
Several types of reactions to drugs and chemicals may precipitate or perpetuate asthmatic relapse. This review focuses on reactions to aspirin and sulfites. Approximately 40 percent of patients with rhinosinusitis, nasal polyps, and asthma and 5 to 10 percent of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service