Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900170

Sigma-Aldrich

Potassium phosphate dibasic trihydrate

Vetec, reagent grade, 99%

Synonym(s):

Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99%
grade:
reagent grade
form:
crystals
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

crystals

pH

8.6-9.4

SMILES string

O.O.O.[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ala A A Al-Subeihi et al.
Toxicology and applied pharmacology, 283(2), 117-126 (2015-01-01)
The present study aims at predicting the level of formation of the ultimate carcinogenic metabolite of methyleugenol, 1'-sulfooxymethyleugenol, in the human population by taking variability in key bioactivation and detoxification reactions into account using Monte Carlo simulations. Depending on the
Karol Kadlec et al.
Talanta, 147, 44-49 (2015-11-26)
The novel approach for hydroxyapatite (HA) surface characterization was proposed. The main aim of this investigation was to estimate surface properties of HA as a biomaterial in real system i.e. in simulated body fluid (SBF). One of the method, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service