V900165
Sodium diethyldithiocarbamate trihydrate
Vetec™, reagent grade
Synonym(s):
Cupral, Diethyldithiocarbamic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein/REAXYS Number:
3920507
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
grade
reagent grade
vapor density
5.9 (vs air)
product line
Vetec™
mp
95-98.5 °C (lit.)
SMILES string
[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]
InChI
1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1
Inchi Key
WWGXHTXOZKVJDN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.
Biochem/physiol Actions
Inhibits induction of macrophage nitric oxide synthase.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mohammad Nasir Uddin et al.
Chemosphere, 90(2), 366-373 (2012-08-28)
A spectrophotometric procedure for the simultaneous determination of zinc(II) and copper(II) in a mixture using diethyldithiocarbamate (DDTC) as ligand has been described. Complexes formed with DDTC was extracted with CHCl(3)/CCl(4) and the absorbance was measured at 435 nm which is
Andreza C Matias et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(1), 349-357 (2012-09-07)
Dithiocarbamates are nitrogen- and sulfur-containing compounds commonly used in pharmacology, medicine and agriculture. The molecular effects of dithiocarbamates on neuronal cell systems are not fully understood, especially in terms of their ability to accumulate copper ions inside the cell. In
Zdenek Skrott et al.
Mini reviews in medicinal chemistry, 12(12), 1184-1192 (2012-08-31)
The idea of "repurposing" of existing drugs provides an effective way to develop and identify new therapies. Disulfiram (Antabuse), a drug commonly used for the treatment of alcoholism, shows promising anticancer activity in both preclinical and clinical studies. In the
Nini Skovgaard et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 303(5), R487-R494 (2012-06-29)
Hypoxic pulmonary vasoconstriction (HPV) is an adaptive response that diverts pulmonary blood flow from poorly ventilated and hypoxic areas of the lung to more well-ventilated parts. This response is important for the local matching of blood perfusion to ventilation and
Bhupesh Sharma et al.
Progress in neuro-psychopharmacology & biological psychiatry, 38(2), 207-215 (2012-04-19)
Dementia of vascular origin or vascular dementia (VaD) is considered as the second commonest form of dementia after Alzheimer's disease (AD). In the last ten years various researchers have reported a strong association of hyperhomocysteinemia (HHcy), hyperlipidemia (HL) and dementia.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service