Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900163

Sigma-Aldrich

Aluminum hydroxide

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)3
CAS Number:
Molecular Weight:
78.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

50.0-57.5% Al (EDTA titration, Al (as Al2O3))

solubility

water: insoluble

density

2.42 g/cm3 at 20 °C

SMILES string

O[Al](O)O

InChI

1S/Al.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

WNROFYMDJYEPJX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaoki Kimura et al.
Water research, 47(6), 2075-2084 (2013-02-21)
Aluminum coagulants are widely used in water treatment plants to remove turbidity and dissolved substances. However, because high aluminum concentrations in treated water are associated with increased turbidity and because aluminum exerts undeniable human health effects, its concentration should be
Micha Phill Grønholm Jepsen et al.
The Journal of infectious diseases, 208(3), 479-488 (2013-04-30)
GMZ2 is a hybrid protein consisting of the N-terminal region of the glutamate-rich protein fused in frame to the C-terminal region of merozoite surface protein 3 (MSP3). GMZ2 formulated in Al(OH)3 has been tested in 3 published phase 1 clinical
Anne Szarewski et al.
The Journal of infectious diseases, 208(9), 1391-1396 (2013-10-05)
Public Health England has reported a decrease of up to 20.8% in new diagnoses of external genital warts (GWs) among women aged <19 years since the national vaccination program with the human papillomavirus (HPV)-16/18 AS04-adjuvanted vaccine began in 2008. A
Wolfgang Hoehenwarter et al.
Molecular & cellular proteomics : MCP, 12(2), 369-380 (2012-11-23)
Mitogen-activated protein kinase (MPK) cascades are important for eukaryotic signal transduction. They convert extracellular stimuli (e.g. some hormones, growth factors, cytokines, microbe- or damage-associated molecular patterns) into intracellular responses while at the same time amplifying the transmitting signal. By doing
Large aqueous aluminum hydroxide molecules.
William H Casey
Chemical reviews, 106(1), 1-16 (2006-01-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service