Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900162

Sigma-Aldrich

Tin(II) oxide

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnO
CAS Number:
Molecular Weight:
134.71
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
assay:
99%
grade:
reagent grade
form:
powder

grade

reagent grade

product line

Vetec

assay

99%

form

powder

SMILES string

O=[SnH2]

InChI

1S/O.Sn

InChI key

QHGNHLZPVBIIPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Tanaka et al.
PloS one, 10(8), e0134386-e0134386 (2015-08-27)
The aortic wall is perfused by the adventitial vasa vasorum (VV). Tissue hypoxia has previously been observed as a manifestation of enlarged abdominal aortic aneurysms (AAAs). We sought to determine whether hypoperfusion of the adventitial VV could develop AAAs. We
Andreas Römpp et al.
Analytical and bioanalytical chemistry, 407(8), 2329-2335 (2015-01-13)
Mass spectrometry imaging has become a popular tool for probing the chemical complexity of biological surfaces. This led to the development of a wide range of instrumentation and preparation protocols. It is thus desirable to evaluate and compare the data
Sean P Hill et al.
The journal of physical chemistry letters, 6(22), 4510-4517 (2015-11-05)
Molecular photon upconversion via triplet-triplet annihilation (TTA-UC), combining two or more low energy photons to generate a higher energy excited state, is an intriguing strategy to surpass the maximum efficiency for a single junction solar cell (<34%). Here, we introduce
Ji-Wook Jang et al.
Nature communications, 6, 7447-7447 (2015-06-17)
Photoelectrochemical (PEC) water splitting promises a solution to the problem of large-scale solar energy storage. However, its development has been impeded by the poor performance of photoanodes, particularly in their capability for photovoltage generation. Many examples employing photovoltaic modules to
Jixian Xu et al.
Nature communications, 6, 7081-7081 (2015-05-09)
Solution-processed planar perovskite devices are highly desirable in a wide variety of optoelectronic applications; however, they are prone to hysteresis and current instabilities. Here we report the first perovskite-PCBM hybrid solid with significantly reduced hysteresis and recombination loss achieved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service