Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900159

Sigma-Aldrich

Potassium sorbate

Vetec, reagent grade, 98%

Synonym(s):

Potassium 2,4-hexadienoate, Sorbic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
Beilstein/REAXYS Number:
5357554
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
98%
grade:
reagent grade

grade

reagent grade

product line

Vetec

assay

98%

SMILES string

[K+].C\C=C\C=C\C([O-])=O

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-An Hwang et al.
Journal of food protection, 78(6), 1154-1160 (2015-06-04)
The surfaces of ready-to-eat meats are susceptible to postprocessing contamination by Listeria monocytogenes. This study quantified the lag-phase durations (LPD) and growth rates (GR) of L. monocytogenes on the surfaces of cooked ham as affected by sorbate solutions of different
Rui Cai et al.
International journal of food microbiology, 214, 145-150 (2015-08-25)
Alicyclobacillus acidoterrestris can survive the pasteurization process, multiply in pasteurized juices and produce guaiacol which causes medicinal or antiseptic off-flavors. Chemical preservatives have the potential to suppress outgrowth of surviving populations during subsequent storage of fruit juices. In the present
Antonio de Castro et al.
Journal of the science of food and agriculture, 95(11), 2222-2227 (2014-10-01)
Alperujo is the paste generated from the two-phase extraction system of olive oil. This wet pomace must be stored for several months and, during this period, the formation of 4-ethyphenol provokes a strong off-odour. The aim of this work was
Giovanna Merchand-Reyes et al.
Journal of medicinal food, 18(11), 1239-1246 (2015-04-14)
Worldwide, the most highly consumed meat is of porcine origin. The production and distribution of swine meat are affected by diverse health matters, such as influenza and diarrhea, which cause head losses and require the use of antibiotics and other
Junjie Wang et al.
Chemistry, an Asian journal, 10(9), 1892-1897 (2015-07-18)
Copper-cobalt bimetal nanoparticles (Cu-Co) have been electrochemically prepared on glassy carbon electrodes (GCEs), which were electrodeposited with conducting polymer nanocomposites of poly(3,4-ethylenedioxythiophene) (PEDOT) doped with carbon nanotubes (CNTs). Owing to their good conductivity, high mechanical strength, and large surface area

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service