V900151
Potassium bicarbonate
Vetec™, reagent grade, 99%
Synonym(s):
Potassium hydrogen carbonate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein/REAXYS Number:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade
reagent grade
product line
Vetec™
assay
99%
form
crystalline powder
crystals or granules
pH
8.2 (10.01 g/L)
SMILES string
[K+].OC([O-])=O
InChI
1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1
Inchi Key
TYJJADVDDVDEDZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bingxing Zhang et al.
Nature communications, 10(1), 2980-2980 (2019-07-07)
Developing highly efficient electrocatalysts based on cheap and earth-abundant metals for CO2 reduction is of great importance. Here we demonstrate that the electrocatalytic activity of manganese-based heterogeneous catalyst can be significantly improved through halogen and nitrogen dual-coordination to modulate the
Pei-Sung Hung et al.
Nanomaterials (Basel, Switzerland), 10(10) (2020-10-01)
We demonstrate the fabrication of free-standing inverse opals with gradient pores via a combination of electrophoresis and electroplating techniques. Our processing scheme starts with the preparation of multilayer colloidal crystals by conducting sequential electrophoresis with polystyrene (PS) microspheres in different
Pei-Sung Hung et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-04)
We demonstrate a water-based synthetic route to fabricate composite inverse opals for simultaneous detection of ascorbic acid (AA), dopamine (DA), and uric acid (UA). Our process involves the conformal deposition of poly(3,4-ethylenedioxythiophene) (PEDOT) and PEDOT/Au on the skeletons of Ni
Judith Buehlmeier et al.
The Journal of clinical endocrinology and metabolism, 97(12), 4789-4797 (2012-10-03)
High sodium chloride (NaCl) intake can induce low-grade metabolic acidosis (LGMA) and may thus influence bone and protein metabolism. We hypothesized that oral potassium bicarbonate (KHCO(3)) supplementation may compensate for NaCl-induced, LGMA-associated bone resorption and protein losses. Eight healthy male
Feng J He et al.
Hypertension (Dallas, Tex. : 1979), 55(3), 681-688 (2010-01-20)
To determine the effects of potassium supplementation on endothelial function, cardiovascular risk factors, and bone turnover and to compare potassium chloride with potassium bicarbonate, we carried out a 12-week randomized, double-blind, placebo-controlled crossover trial in 42 individuals with untreated mildly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service