Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900148

Sigma-Aldrich

Tetrabutylammonium hydrogensulfate

Vetec, reagent grade, 97%

Synonym(s):

Tetrabutylammonium bisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

169-171 °C (lit.)

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

348.1 °F - closed cup

flash_point_c

175.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of
Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind
Toshihiro Kimura et al.
Inorganic chemistry, 51(23), 13001-13008 (2012-11-15)
A simple monomeric tungstate, TBA(2)[WO(4)] (I, TBA = tetra-n-butylammonium), could act as an efficient homogeneous catalyst for chemical fixation of CO(2) with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Various kinds of structurally diverse 2-aminobenzonitriles could be converted into the corresponding quinazoline-2,4(1H,3H)-diones in high
K M Frankowski et al.
Journal of dairy science, 97(9), 5356-5370 (2014-07-16)
Many food companies are trying to limit the amount of sodium in their products. Permeate, the liquid remaining after whey or milk is ultrafiltered, has been suggested as a salt substitute. The objective of this study was to determine the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service