Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900139

Sigma-Aldrich

Tetrapropylammonium hydroxide solution

Vetec, reagent grade

Synonym(s):

TPAOH solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
203.36
Beilstein/REAXYS Number:
4207983
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

SMILES string

[OH-].CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.H2O/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H2/q+1;/p-1

InChI key

LPSKDVINWQNWFE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth I J Amos et al.
Angewandte Chemie (International ed. in English), 53(42), 11275-11279 (2014-08-30)
A robust catalyst for the selective dehydrogenation of formic acid to liberate hydrogen gas has been designed computationally, and also successfully demonstrated experimentally. This is the first such catalyst not based on transition metals, and it exhibits very encouraging performance.
Weiwei Yuan et al.
Journal of colloid and interface science, 462, 191-199 (2015-10-12)
Hierarchically porous TS-1/modified-diatomite composites with high removal efficiency for methylene blue (MB) were prepared via a facile in situ hydrothermal route. The surface charge state of the diatomite was modified to enhance the electrostatic interactions, followed by in situ hydrothermal
Juna Khang et al.
Talanta, 147, 177-183 (2015-11-26)
Cost-effective and easy-to-use biosensor was developed for the rapid quantification and monitoring of Escherichia (E.) Coli O157:H7 in sample using E. Coli O157:H7 aptamer, graphene oxide (GO)/iron nanocomposites, and guanine chemiluminescence detection. E. Coli O157:H7 aptamer-conjugated 6-carboxyfluorescein (6-FAM) with excellent
Timothy Cha et al.
Biosensors & bioelectronics, 62, 31-37 (2014-06-29)
Using guanine (G)-rich DNA aptamer-conjugated 6-carboxyfluorescein (6-FAM) capable of rapidly capturing prostate specific antigen (PSA) in human serum, cost-effective and simple biosensor with guanine chemiluminescence detection was developed for early diagnosis of prostate cancer. Free G-rich DNA aptamer-conjugated 6-FAM emits
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service