Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900131

Sigma-Aldrich

CDI

Vetec, reagent grade, 97%

Synonym(s):

1,1′-Carbonyldiimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4O
CAS Number:
Molecular Weight:
162.15
Beilstein/REAXYS Number:
6826
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

117-122 °C (lit.)

SMILES string

O=C(n1ccnc1)n2ccnc2

InChI

1S/C7H6N4O/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6H

InChI key

PFKFTWBEEFSNDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CDI can be used:
  • To decarboxylate malonic acid derivatives to yield carbonyl imidazole moiety, which can further be reacted with different nucleophiles.[1]
  • To activate cellulose membranes, which are used in immunosensor applications.[2]
  • To functionalize Fe3O4-SiO2 core-shell nanoparticles for bio-applications.[3]
  • To prepare syn- and anti-1,2-imidazolylpropylamines by reacting with syn- and anti-1,2-amino alcohols.[4]
  • In the preparation of N-substituted imidazole derivatives, which are further used to prepare new ionic liquids.[5]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regio-and Stereospecific Syntheses of syn-and anti-1, 2-Imidazolylpropylamines from the Reaction of 1, 1′-Carbonyldiimidazole with syn-and anti-1, 2-Amino Alcohols
Mulvihill MJ, et al.
The Journal of Organic Chemistry, 69(15), 5124-5127 (2004)
Synthesis and surface functionalization of Fe3O4-SiO2 core-shell nanoparticles with 3-glycidoxypropyltrimethoxysilane and 1, 1′-carbonyldiimidazole for bio-applications
Ta TKH, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 504(2), 376-383 (2016)
1, 1′-Carbonyldiimidazole mediates the synthesis of N-substituted imidazole derivatives from Morita--Baylis--Hillman adducts
Rodrigues Jr MT, et al.
Tetrahedron Letters, 55(1), 180-183 (2014)
Daniela Stöllner et al.
Analytical biochemistry, 304(2), 157-165 (2002-05-16)
Methods for the activation of a cellulose dialysis membrane for immunosensor applications have been developed. For activation two reagents, 1,1'-carbonyldiimidazole (CDI) and 1-cyano-4-dimethylaminopyridinium tetrafluoroborate (CDAP), were compared with respect to the coupling efficiency for glucose oxidase (GOx) and 1,8-diamino-2,6-dioxaoctane. The
Frank Münch et al.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery, 48(2), 236-243 (2014-11-22)
Experience regarding warm blood cardioplegia according to Calafiore results from its broad use in adult patients. In this experimental study, tepid (28°C) modified full blood cardioplegia (MBC) was adopted for paediatric use and compared with cold crystalloid cardioplegia (CCC). Twenty

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service