V900124
Lithium carbonate
Vetec™, reagent grade, 98%
Synonym(s):
Carbolithium, Carbonic acid lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein/REAXYS Number:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
powder
grade
reagent grade
product line
Vetec™
assay
98%
form
powder
pH
9.0-11.0 (1 g/L)
mp
618 °C (lit.)
SMILES string
[Li+].[Li+].[O-]C([O-])=O
InChI
1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
Inchi Key
XGZVUEUWXADBQD-UHFFFAOYSA-L
Gene Information
human ... GSK3A(2931) , GSK3B(2932) , IMPA1(3612)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Andrew A Nierenberg et al.
The American journal of psychiatry, 170(1), 102-110 (2013-01-05)
Lithium salts, once the mainstay of therapy for bipolar disorder, have tolerability issues at a higher dosage that often limit adherence. The authors investigated the comparative effectiveness of more tolerable dosages of lithium as part of optimized personalized treatment (OPT).
Harm-Pieter Spaans et al.
The Journal of clinical psychiatry, 74(11), e1029-e1036 (2013-12-18)
To compare the efficacy and cognitive side effects of high-dose unilateral brief pulse electroconvulsive therapy (ECT) with those of high-dose unilateral ultrabrief pulse ECT in the treatment of major depression. From April 2007 until March 2011, we conducted a prospective
Tom Bschor et al.
Journal of clinical psychopharmacology, 33(1), 38-44 (2013-01-02)
Studies of the 1970s and 1980s showed lithium monotherapy to be an effective treatment of acute unipolar major depressive disorder (MDD) and hence as a potential alternative to monoaminergic antidepressants.The objective was to conduct the first comparison of a lithium
Andrea Contestabile et al.
The Journal of clinical investigation, 123(1), 348-361 (2012-12-04)
Down syndrome (DS) patients exhibit abnormalities of hippocampal-dependent explicit memory, a feature that is replicated in relevant mouse models of the disease. Adult hippocampal neurogenesis, which is impaired in DS and other neuropsychiatric diseases, plays a key role in hippocampal
K E Morrison et al.
The Lancet. Neurology, 12(4), 339-345 (2013-03-05)
Lithium has neuroprotective effects in cell and animal models of amyotrophic lateral sclerosis (ALS), and a small pilot study in patients with ALS showed a significant effect of lithium on survival. We aimed to assess whether lithium improves survival in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service