Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900120

Sigma-Aldrich

Stearic acid

Vetec, reagent grade, 94%

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein/REAXYS Number:
608585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

1 mmHg ( 173.7 °C)

product line

Vetec

assay

94%

form

powder or flakes

bp

361 °C (lit.)

mp

67-72 °C (lit.)

solubility

chloroform: 10%, clear to very slightly hazy, colorless to faintly yellow

density

0.845 g/cm3

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulla Hoppu et al.
Journal of pediatric gastroenterology and nutrition, 41(3), 335-338 (2005-09-01)
Breast milk fatty acids may have immunomodulatory properties related to the development of atopic disease. The aim of this study was to assess the impact of the breast milk fatty acid composition on the development of atopic dermatitis (AD) in
Qian Wan et al.
Nanoscale, 5(2), 744-752 (2012-12-12)
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron
T A Doran et al.
American journal of obstetrics and gynecology, 133(3), 302-307 (1979-02-01)
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples
Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting
A M Turpeinen et al.
Arteriosclerosis, thrombosis, and vascular biology, 18(2), 316-322 (1998-03-04)
The effects of stearic acid (C18:0) and trans-fatty acids (trans-FAs) on measures of platelet function and prostacyclin (PGI2) production are poorly understood in humans. In this controlled dietary study, platelet function and endothelial PGI2 production were studied in healthy humans

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service