Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900117

Sigma-Aldrich

Cobalt(II) carbonate hydrate

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCO3·xH2O
CAS Number:
Molecular Weight:
118.94 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for synthesis
reagent grade
synthesis grade
form:
powder
Pricing and availability is not currently available.

grade

for synthesis
reagent grade
synthesis grade

product line

Vetec

form

powder

concentration

43.0-47.0% Co (EDTA titration)

density

4.13 g/mL at 25 °C (lit.)

SMILES string

O.[Co++].[O-]C([O-])=O

InChI

1S/CH2O3.Co.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-2

InChI key

JLAIPADPFFTYLP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service