Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900113

Sigma-Aldrich

Potassium benzoate

Vetec, reagent grade, 98%

Synonym(s):

Benzoic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOK
CAS Number:
Molecular Weight:
160.21
Beilstein/REAXYS Number:
3719165
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

SMILES string

[K+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.K/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

XAEFZNCEHLXOMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dajie Lin et al.
Biosensors & bioelectronics, 45, 195-200 (2013-03-19)
An amplification strategy for signal tracing was developed by introducing a host-guest binding reaction into the assembly process of gold nanorods (AuNRs) superstructure. The amplification pathway firstly used a thio-β-cyclodextrin (SH-β-CD) functionalized gold nanoparticles to label signal antibody, and then
Philipp Ottiger et al.
The Journal of chemical physics, 137(20), 204303-204303 (2012-12-05)
The benzoic acid dimer, (BZA)(2), is a paradigmatic symmetric hydrogen bonded dimer with two strong antiparallel hydrogen bonds. The excitonic S(1)/S(2) state splitting and coherent electronic energy transfer within supersonically cooled (BZA)(2) and its (13)C-, d(1) -, d(2) -, and
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Miyuki Kusajima et al.
Bioscience, biotechnology, and biochemistry, 76(12), 2301-2306 (2012-12-12)
Systemic acquired resistance (SAR) is a potent innate immunity system in plants and has been used in rice fields. Development of SAR, involving priming, is achieved by activation of salicylic acid (SA)-mediated pathway. To determine whether heat shock (HS) treatment
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service