V900109
5-Sulfosalicylic acid dihydrate
Vetec™, reagent grade, 98%
Synonym(s):
2-Hydroxy-5-sulfobenzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein/REAXYS Number:
650741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
grade
reagent grade
product line
Vetec™
assay
98%
mp
105-110 °C (lit.)
SMILES string
[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O
InChI
1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2
InChI key
BHDKTFQBRFWJKR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Vázquez et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 74, 349-359 (2014-12-03)
The mercurial forms [inorganic divalent mercury, Hg(II) and methylmercury, CH3Hg] produce neurological and immune effects as well as hematological and renal alterations. The main route of exposure is through the diet. Consequently, the gastrointestinal mucosa is exposed to these mercurial
Yu Zhu et al.
Molecular pharmaceutics, 12(2), 332-341 (2014-08-26)
Polycations are explored as carriers to deliver therapeutic nucleic acids. Polycations are conventionally pharmacological inert with the sole function of delivering therapeutic cargo. This study reports synthesis of a self-immolative polycation (DSS-BEN) based on a polyamine analogue drug N(1),N(11)-bisethylnorspermine (BENSpm).
Swati Naphade et al.
Stem cells (Dayton, Ohio), 33(1), 301-309 (2014-09-05)
Despite controversies on the potential of hematopoietic stem cells (HSCs) to promote tissue repair, we previously showed that HSC transplantation could correct cystinosis, a multisystemic lysosomal storage disease, caused by a defective lysosomal membrane cystine transporter, cystinosin (CTNS gene). Addressing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service