Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900104

Sigma-Aldrich

Potassium citrate tribasic monohydrate

Vetec, reagent grade, 98%

Synonym(s):

Citric acid tripotassium salt, Tripotassium citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOK)(CH2COOK)2 · H2O
CAS Number:
Molecular Weight:
324.41
Beilstein/REAXYS Number:
3924344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
98%
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

275 °C (dec.) (lit.)

SMILES string

O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3K.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

PJAHUDTUZRZBKM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun Gopalakrishnan et al.
Clinical and experimental nephrology, 15(5), 745-748 (2011-06-01)
Fanconi-Bickel syndrome is a rare autosomal recessive disorder characterized by hepatorenal glycogen accumulation, proximal renal tubular dysfunction and impaired utilization of glucose and galactose. Most cases have been reported from Europe, Japan, Turkey and the Mediterranean belt. We report a
Zeynep Yarbaşi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1304-1307 (2011-06-07)
An EPR and optical studies of VO2+ doped potassium dihydrogen citrate (PDHC) single crystals have been carried out at room temperature. It crystallizes in triclinic symmetry with the unit cell dimensions: a=11.343Å, b=13.078Å, c=6.272Å, α=89.79°, β=94.36°, γ=104.2°. The angular variation
Pierre Cochat et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 27(5), 1729-1736 (2012-05-02)
Primary hyperoxaluria Type 1 is a rare autosomal recessive inborn error of glyoxylate metabolism, caused by a deficiency of the liver-specific enzyme alanine:glyoxylate aminotransferase. The disorder results in overproduction and excessive urinary excretion of oxalate, causing recurrent urolithiasis and nephrocalcinosis.
Maria Goretti Moreira Guimarães Penido et al.
Pediatric nephrology (Berlin, Germany), 27(1), 123-130 (2011-07-23)
Children with idiopathic hypercalciuria (IH) may have a reduced bone mineral density (BMD), which could impact on bone health in adulthood. There is currently no strong evidence for a preferred treatment of such children. The aim of our study was
Astrid Starke et al.
Clinical journal of the American Society of Nephrology : CJASN, 7(9), 1461-1472 (2012-07-10)
Acidosis and transplantation are associated with increased risk of bone disturbances. This study aimed to assess bone morphology and metabolism in acidotic patients with a renal graft, and to ameliorate bone characteristics by restoration of acid/base homeostasis with potassium citrate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service