Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900102

Sigma-Aldrich

Sodium succinate dibasic hexahydrate

Vetec, reagent grade, 98%

Synonym(s):

Butanedioic acid disodium salt, Succinic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2COONa · 6H2O
CAS Number:
Molecular Weight:
270.14
Beilstein/REAXYS Number:
3921044
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
98%
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

InChI

1S/C4H6O4.2Na.6H2O/c5-3(6)1-2-4(7)8;;;;;;;;/h1-2H2,(H,5,6)(H,7,8);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

ZBTUYCUNQBRXOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Kimura et al.
Cancer science, 104(8), 1083-1090 (2013-05-18)
Pancreatic cancer has a poor prognosis after complete macroscopic resection combined with chemotherapy. Even after neoadjuvant chemotherapy, R0 resection is often not possible. Moreover, current imaging techniques cannot reliably distinguish viable cancer cells from scar tissue at the resectional margin.
Ilya B Zavodnik et al.
Life sciences, 92(23), 1110-1117 (2013-05-07)
The present study was designed for further evaluation of the biochemical mechanism of hepatic mitochondrial dysfunction under oxidative damages induced by organic hydroperoxide, tert-butyl hydroperoxide (tBHP), for estimation of the molecular targets impaired during oxidative stress, and for investigation of
Hiroshi Yoneyama et al.
Anti-cancer drugs, 26(1), 90-100 (2014-09-05)
The efficacy of gemcitabine (GEM), a standard treatment agent for pancreatic cancer, is insufficient because of primary or acquired resistance to this drug. Patients with tumors intrinsically sensitive to GEM gradually acquire resistance and require a shift to second agents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service