Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900093

Sigma-Aldrich

Cobalt(II) acetate tetrahydrate

Vetec, reagent grade

Synonym(s):

Cobaltous acetate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Co · 4H2O
CAS Number:
Molecular Weight:
249.08
Beilstein/REAXYS Number:
3731397
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
grade:
reagent grade
form:
powder or crystals
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder or crystals

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Co]OC(C)=O

InChI

1S/2C2H4O2.Co.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

ZBYYWKJVSFHYJL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijayan Thamilarasan et al.
European journal of medicinal chemistry, 89, 266-278 (2014-12-03)
Metal complexes of the type Mn(bpy)2(N3)2 (1), Co(bpy)2(N3)2·3H2O (2) and Zn2(bpy)2(N3)4 (3) (Where bpy = 2,2-bipyridine) have been synthesized and characterized by elemental analysis and spectral (FT-IR, UV-vis) studies. The structure of complexes (1-3) have been determined by single crystal X-ray diffraction
Kyu-Nam Jung et al.
Scientific reports, 5, 7665-7665 (2015-01-08)
Rechargeable metal-air batteries are considered a promising energy storage solution owing to their high theoretical energy density. The major obstacles to realising this technology include the slow kinetics of oxygen reduction and evolution on the cathode (air electrode) upon battery
Yun-Jung Kim et al.
ChemSusChem, 8(15), 2496-2502 (2015-07-17)
The problem of high charge polarization is one of the most significant challenges in current nonaqueous Li-O2 batteries. The development of an electrode for the oxygen evolution reaction (OER) at reduced overpotential is thus essential. Here, we suggest a binder-free

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service