Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900087

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

Vetec, reagent grade, 98%

Synonym(s):

4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, p-Toluenesulfonic acid, PTSA monohydrate, TsOH monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein/REAXYS Number:
3568023
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.9 (vs air)

product line

Vetec

assay

98%

mp

103-106 °C (lit.)

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild C-H halogenation of anilides and the isolation of an unusual palladium(I)-palladium(II) species.
Robin B Bedford et al.
Angewandte Chemie (International ed. in English), 50(24), 5524-5527 (2011-05-14)
Hans H Adomat et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 902, 84-95 (2012-07-24)
Androgens are key mediators of prostate development and function, a role that extends to the development of prostate diseases such as benign prostatic hyperplasia (BPH) and prostate cancer. In prostate, DHT is the major androgen and reduction and glucuronidation are
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the
Pei Liu et al.
Organic & biomolecular chemistry, 10(24), 4696-4698 (2012-05-18)
A convenient one-pot Mannich-type-cyclization-oxidation tandem process has been developed for the synthesis of 1,3,5-trisubstituted pyrazoles derivatives from aldehydes, hydrazines and alkynes using p-toluenesulfonic acid monohydrate (PTSA) as a multifunctional catalyst. This method provides a flexible and rapid route to 1,3,5-trisubstituted
Huai-Long Teng et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(40), 12614-12618 (2012-08-29)
Spiro(γ-butyrolactam-γ-butyrolactone): a route to enantioenriched spiro(γ-butyrolactam-γ-butyrolactone) compounds, a valuable motif for drug discovery, was developed by use of a highly efficient copper(I)/TF-BiphamPhos-catalyzed tandem Michael addition-elimination of homoserine lactone derived cyclic imino esters with Morita-Baylis-Hillman (MBH) bromides, followed by treatment with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service