Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900082

Sigma-Aldrich

Potassium carbonate sesquihydrate

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3 · 1.5H2O
CAS Number:
Molecular Weight:
165.23
EC Number:
UNSPSC Code:
12352300
assay:
99%
grade:
reagent grade
form:
chunks or granules
crystalline powder
powder or crystals

grade

reagent grade

product line

Vetec

assay

99%

form

chunks or granules
crystalline powder
powder or crystals

SMILES string

O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service