Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900064

Sigma-Aldrich

Formamide

Vetec, reagent grade, 98%

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein/REAXYS Number:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
grade:
reagent grade
assay:
98%
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

grade

reagent grade

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

product line

Vetec

assay

98%

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 % (lit.)

technique(s)

hybridization: suitable

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

target_organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Chen et al.
Prenatal diagnosis, 35(6), 534-543 (2015-02-03)
To develop a single-tube multi-marker assay for improved preimplantation genetic diagnosis (PGD) of deletional and/or non-deletional Hb Bart's hydrops fetalis syndrome, providing haplotype confirmation of deletional status, and maximization of linkage informativity. We performed in silico mining to identify novel
Yue Li et al.
RNA (New York, N.Y.), 21(8), 1511-1518 (2015-06-21)
N(6)-methyladenosine (m(6)A), the most abundant internal RNA modification, functions in diverse biological processes, including regulation of embryonic stem cell self-renewal and differentiation. As yet, methods to detect m(6)A in the transcriptome rely on the availability and quality of an m(6)A
Jing Wang et al.
The journal of physical chemistry. B, 117(8), 2314-2320 (2013-01-26)
A step-by-step mechanistic pathway following the transformation of formamide to purine through a five-membered ring intermediate has been explored by density functional theory computations. The highlight of the mechanistic route detailed here is that the proposed pathway represents the simplest
A Touati et al.
Journal of clinical microbiology, 53(10), 3182-3194 (2015-07-24)
Molecular typing of Mycoplasma pneumoniae is an important tool for identifying grouped cases and investigating outbreaks. In the present study, we developed a new genotyping method based on single nucleotide polymorphisms (SNPs) selected from the whole-genome sequencing of eight M.
J Veenemans et al.
Journal of clinical microbiology, 52(7), 2454-2460 (2014-05-03)
Next-generation sequencing (NGS) has the potential to provide typing results and detect resistance genes in a single assay, thus guiding timely treatment decisions and allowing rapid tracking of transmission of resistant clones. We evaluated the performance of a new NGS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service