Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900059

Sigma-Aldrich

Cobalt(II) chloride

Vetec, reagent grade

Synonym(s):

Cobalt dichloride, Cobalt muriate, Cobaltous chloride, Dichlorocobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2
CAS Number:
Molecular Weight:
129.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

40 mmHg ( 0 °C)

product line

Vetec

form

powder

concentration

43.7-47.0% Co (complexometric)

mp

724 °C (lit.)

SMILES string

Cl[Co]Cl

InChI

1S/2ClH.Co/h2*1H;/q;;+2/p-2

InChI key

GVPFVAHMJGGAJG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongming Teng et al.
Marine drugs, 13(6), 3514-3530 (2015-06-06)
Metastasis, the greatest clinical challenge associated with cancer, is closely connected to multiple biological processes, including invasion and adhesion. The hypoxic environment in tumors is an important factor that causes tumor metastasis by activating HIF-1α. Fucoidan, extracted from brown algae
Katrin Orlowski et al.
PloS one, 7(12), e51476-e51476 (2012-12-20)
Tumor hypoxia is one of the most important parameters that determines treatment sensitivity and is mainly due to insufficient tumor angiogenesis. However, the local oxygen concentration in a tumor can also be shifted in response to different treatment modalities such
Nobukazu Miyamoto et al.
Stroke, 44(9), 2573-2578 (2013-07-25)
Aging is one of the major risk factors for white matter injury in cerebrovascular disease. However, the effects of age on the mechanisms of injury/repair in white matter remain to be fully elucidated. Here, we ask whether, compared with young
Thomas Naves et al.
Biochemical pharmacology, 85(8), 1153-1161 (2013-02-06)
Solid tumors like neuroblastoma exhibit hypoxic areas, which can lead both to cell death or aggressiveness increase. Hypoxia is a known stress able to induce stabilization of p53, implicated in cell fate regulation. Recently, p53 appeared to be involved in
Jérémy Denizot et al.
Gut, 64(3), 428-437 (2014-06-06)
Adherent-invasive Escherichia coli (AIEC) are abnormally predominant on Crohn's disease (CD) ileal mucosa. AIEC strains adhere to enterocytes via interaction between type 1 pili and CEACAM6 receptors abnormally expressed on CD ileal mucosa, leading to gut inflammation. We analysed whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service