Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900054

Sigma-Aldrich

Sodium iodide

Vetec, reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
83.8-85.4% iodine (I) basis (silver nitrate titration)
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

vapor density

>1 (vs air)

product line

Vetec

assay

83.8-85.4% iodine (I) basis (silver nitrate titration)

form

powder

pH

6.0-9.0 (20 °C, 50%)

mp

661 °C (lit.)

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

1203.8 °F - closed cup

flash_point_c

651 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brad A Bauer et al.
Physical chemistry chemical physics : PCCP, 14(6), 1892-1906 (2012-01-11)
We present results from all-atom molecular dynamics simulations of large-scale hydrophobic plates solvated in NaCl and NaI salt solutions. As observed in studies of ions at the air-water interface, the density of iodide near the water-plate interface is significantly enhanced
Anke de Vries et al.
Investigative radiology, 50(4), 297-304 (2015-01-01)
The objective of this study was to investigate the feasibility and the accuracy of spectral computed tomography (spectral CT) to determine the tissue concentrations and localization of high-attenuation, iodine-based contrast agents in mice. Iodine tissue concentrations determined with spectral CT
Caroline Serrano-Nascimento et al.
Molecular and cellular endocrinology, 349(2), 154-161 (2011-10-18)
Iodide excess acutely downregulates NIS mRNA expression, as already demonstrated. PCCl3 cells treated or not with NaI, NaI+NaClO(4) or NaI+Methimazole, for 30 min to 24 h, were used to further explore how iodide reduces NIS gene expression. NIS mRNA expression
Lan-Tao Li et al.
Chemical communications (Cambridge, England), 48(42), 5187-5189 (2012-04-20)
nBu(4)NI-catalyzed C3-selective formylation of N-H and N-substituted indoles by using N-methylaniline as a formylating reagent was first successfully demonstrated.
Dafu Yu et al.
Hormones and behavior, 69, 106-115 (2015-01-28)
Thyroid hormone disorders have long been linked to depression, but the causal relationship between them remains controversial. To address this question, we established rat models of hypothyroidism using (131)iodine ((131)I) and hyperthyroidism using levothyroxine (LT4). Serum free thyroxine (FT4) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service