Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900043

Sigma-Aldrich

Calcium sulfate

Vetec, reagent grade

Synonym(s):

Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

concentration

29.0-29.9% Ca (EDTA titration)

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark V Thomas et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 88(2), 597-610 (2008-11-26)
Calcium sulfate (CS) has enjoyed a longer history of clinical use than most currently available biomaterials. It is well-tolerated when used to fill bone defects and undergoes rapid and complete resorption without eliciting a significant inflammatory response. The raw material
Kathryn A Dooley et al.
The Analyst, 138(17), 4838-4848 (2013-06-27)
In situ chemical imaging techniques are being developed to provide information on the spatial distribution of artists' pigments used in polychrome works of art such as paintings. The new methods include reflectance imaging spectroscopy and X-ray fluorescence mapping. Results from
Joon Im et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 434-442 (2014-04-08)
The aim of this study was to compare the virtual and manual tooth setups with digital and plaster models in extraction cases by measuring various occlusal parameters and applying the American Board of Orthodontics objective grading system. Linear intra-arch and
Nathaniel Huebsch et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9762-9767 (2014-06-26)
Biological systems are exquisitely sensitive to the location and timing of physiologic cues and drugs. This spatiotemporal sensitivity presents opportunities for developing new therapeutic approaches. Polymer-based delivery systems are used extensively for attaining localized, sustained release of bioactive molecules. However
Aletta Hazeveld et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(1), 108-115 (2014-01-01)
Rapid prototyping is a fast-developing technique that might play a significant role in the eventual replacement of plaster dental models. The aim of this study was to investigate the accuracy and reproducibility of physical dental models reconstructed from digital data

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service