Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900040

Sigma-Aldrich

Potassium sulfate

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

powder

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined
C C Constantinoiu et al.
Poultry science, 86(9), 1910-1914 (2007-08-21)
Immunoglobulin Y is different from most of the other immunoglobulins because it does not bind protein A or protein G. Thiophilic gel chromatography has been successfully used to purify IgY from chicken egg yolk, but the technology has not previously
Jingjing Zheng et al.
Bioresource technology, 110, 224-231 (2012-02-15)
This study investigated the preparation and characteristics of protein-coated microcrystals (PCMCs) from Pseudomonas cepacia lipase (PS) and K(2)SO(4), and their application in biodiesel synthesis, via single factorial experiments and response surface methodology (RSM), the optimized PCMC-PS exhibited high activity and
Xinhua Xu et al.
Journal of hazardous materials, 158(2-3), 410-416 (2008-03-21)
The aqueous phase oxidation of gaseous elemental mercury (Hg(0)) by potassium persulfate (K(2)S(2)O(8), KPS) catalyzed by Ag(+) and Cu(2+) was investigated using a glass bubble column reactor. Concentrations of gaseous Hg(0) and aqueous Hg(2+) were measured by cold vapor generation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service