V900035
Ammonium sodium phosphate dibasic tetrahydrate
Vetec™, reagent grade, 98%
Synonym(s):
sec-Sodium ammonium phosphate, Ammonium sodium hydrogen phosphate, Sodium ammonium hydrogen phosphate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NaNH4HPO4 · 4H2O
CAS Number:
Molecular Weight:
209.07
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
crystals
grade
reagent grade
product line
Vetec™
assay
98%
form
crystals
SMILES string
[NH4+].[Na+].OP([O-])([O-])=O
InChI
1S/H3N.Na.H3O4P.4H2O/c;;1-5(2,3)4;;;;/h1H3;;(H3,1,2,3,4);4*1H2/q;+1;;;;;/p-1
InChI key
IQTQISLCLWJRPM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Keiko Inami et al.
Bioorganic & medicinal chemistry, 23(13), 3297-3302 (2015-05-16)
N-Nitroso-N-methylurea (NMU) is a potent carcinogen and suspected as a cause of human cancer. In this study, mutagenic NMU was detected by HPLC after the transnitrosation of non-mutagenic N-nitrosoproline (NP) to N-methylurea in the presence of thiourea (TU) under acidic
Toine F H Bovee et al.
Mutation research, 778, 1-10 (2015-05-30)
It has previously been demonstrated by others that acetone extracts of Senecio jacobaea (syn. Jacobaea vulgaris, common or tansy ragwort) test positive in the Salmonella/microsome mutagenicity test (Ames test). Pyrrolizidine alkaloids (PAs) are thought to be responsible for these mutagenic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service