Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900027

Sigma-Aldrich

Bismuth(III) chloride

Vetec, reagent grade

Synonym(s):

Bismuth trichloride, Trichlorobismuth

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiCl3
CAS Number:
Molecular Weight:
315.34
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

chunks or granules
powder or crystals

reaction suitability

reagent type: catalyst
core: bismuth

bp

447 °C (lit.)

mp

230-232 °C (lit.)

SMILES string

Cl[Bi](Cl)Cl

InChI

1S/Bi.3ClH/h;3*1H/q+3;;;/p-3

InChI key

JHXKRIRFYBPWGE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunqing Zhu et al.
ACS nano, 8(5), 5022-5031 (2014-04-09)
Homopolymers have been considered as a nonideal building block for creating well-defined nanostructures due to their fuzzy boundary between hydrophobic and hydrophilic moieties. However, this unique fuzzy boundary may provide some opportunities for fabricating functional nanomaterials. Presented in this paper

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service