Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900015

Sigma-Aldrich

NMP

Vetec, reagent grade, 98%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein/REAXYS Number:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
reagent grade
assay:
98%
bp:
202 °C (lit.)
81-82 °C/10 mmHg (lit.)
vapor pressure:
0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Product Name

1-Methyl-2-pyrrolidinone, Vetec, reagent grade, 98%

grade

reagent grade

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

product line

Vetec

assay

98%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

dilution

(for analytical testing)

refractive index

n20/D 1.47 (lit.)

pH

7.7-8

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B A Jönsson et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(2), 351-357 (1997-07-04)
A method for simultaneous determination of 5-hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide in urine is described. These compounds are metabolites of N-methyl-2-pyrrolidone, a powerful and widely used organic solvent. 5-Hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide were purified from urine by adsorption to a C8 solid-phase extraction
Laura Vogelaar et al.
Small (Weinheim an der Bergstrasse, Germany), 1(6), 645-655 (2006-12-29)
Phase separation micromolding (PSmicroM) is a versatile microfabrication technique that can be used to structure a very broad range of polymers, including block copolymers and biodegradable and conductive polymers without the need for clean-room facilities. By incorporating a subsequent process
Hsiao-Chun Wang et al.
European journal of medicinal chemistry, 84, 312-334 (2014-07-19)
Bioisosteric replacement of acylureido moiety in 6-acylureido-3-pyrrolylmethylidene-2-oxoindoline derivatives resulted in a series of malonamido derivatives with indolin-2-one scaffold (11-14). Further conformational restrictions of the malonamido moiety led to 2-oxo-1,2-dihydropyridine (21-25) or a 4-oxo-1,4-dihydropyridine derivatives (31-36). 4-Oxo-1,4-dihydropyridine derivatives were more potent
Ajit Dhananjay Jagtap et al.
European journal of medicinal chemistry, 85, 268-288 (2014-08-05)
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50 = 0.4 nM and 0.5 nM, respectively).
Yichao Wang et al.
Nano letters, 15(2), 883-890 (2015-01-07)
The exhibition of plasmon resonances in two-dimensional (2D) semiconductor compounds is desirable for many applications. Here, by electrochemically intercalating lithium into 2D molybdenum disulfide (MoS2) nanoflakes, plasmon resonances in the visible and near UV wavelength ranges are achieved. These plasmon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service